missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diphenylethane, 98+%

CAS: 103-29-7 | C14H14 | 182.27 g/mol

Supplier:  Thermo Scientific Chemicals A1461722

 Afficher plus de versions de ce produit

Numéro de catalogue. AAA1461722


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

1,2-Diphenylethane is used as solvent for nitro fibre and in synthesis of other organic chemical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diphenylethane is used as solvent for nitro fibre and in synthesis of other organic chemical products.

Solubility
Soluble in ether, chloroform

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Spécifications

Spécifications

1,2-Diphenylethane
50°C to 53°C
284°C
C14H14
100 g
14,1198
Soluble in ether, chloroform
C(CC1=CC=CC=C1)C1=CC=CC=C1
182.27
CHEBI:34047
≥98%
103-29-7
1.013
>110°C (230°F)
MFCD00004796
508068
1,2-diphenylethane, bibenzyl, dibenzyl, s-diphenylethane, 2-phenylethyl benzene, 1,2-dihydrostilbene, dibenzil, sym-diphenylethane, dihydrostilbene, benzene, 1,1'-1,2-ethanediyl bis
QWUWMCYKGHVNAV-UHFFFAOYSA-N
2-phenylethylbenzene
7647
182.27

Identifiants chimiques

103-29-7
182.27
QWUWMCYKGHVNAV-UHFFFAOYSA-N
7647
2-phenylethylbenzene
C14H14
MFCD00004796
1,2-diphenylethane, bibenzyl, dibenzyl, s-diphenylethane, 2-phenylethyl benzene, 1,2-dihydrostilbene, dibenzil, sym-diphenylethane, dihydrostilbene, benzene, 1,1'-1,2-ethanediyl bis
CHEBI:34047
C(CC1=CC=CC=C1)C1=CC=CC=C1
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'einecsNumber' : 203-096-4

missing translation for 'rtecsNumber' : DT4375000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only