missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane (Certified ACS), Fisher Chemical™

$1320.27 - $1320.27

Identifiants chimiques

CAS 107-06-2
Molecular Formula C2H4Cl2
Molecular Weight (g/mol) 98.95
MDL Number MFCD00000963
InChI Key WSLDOOZREJYCGB-UHFFFAOYSA-N
Synonym ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
PubChem CID 11
ChEBI CHEBI:27789
IUPAC Name 1,2-dichloroethane
SMILES ClCCCl
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
E175-4
FDS Accessoires Afficher les documents
Fisher Chemical
E1754
4 L Glass Bottle
chaque for $1,320.27
caisse de 4 chaque for $5,281.08
Il en reste null
 
Description

Description

Spécifications

Identifiants chimiques

107-06-2
98.95
WSLDOOZREJYCGB-UHFFFAOYSA-N
11
1,2-dichloroethane
C2H4Cl2
MFCD00000963
ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
CHEBI:27789
ClCCCl

Spécifications

-35°C
Colorless
81°C
13°C (55.4°F)
Glass Bottle
0.002% max.
0.0003mEq/g max.
MFCD00000963
0.8 mPa/s at 20°C
0.03% max.
WSLDOOZREJYCGB-UHFFFAOYSA-N
1,2-dichloroethane
11
98.96
Certified ACS Reagent
107-06-2
1.250g/cm³
4 L
Pass Test
Liquid
≥99 %
C2H4Cl2
65mmHg at 29°C
1184
ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
ClCCCl
98.95
CHEBI:27789
≥99%
1, 2-Dichloroethane
Videos
Sécurité et manipulation

Sécurité et manipulation

DANGER!

Emergency Overview
Flammable liquid and vapor. Harmful if swallowed. Irritating to eyes, respiratory system and skin. May cause central nervous system effects. May cause adverse liver effects. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0

missing translation for 'dotInformation' : DOT Class 3, : Flammable Liquid

FDS
missing translation for 'documents'

missing translation for 'documents'