Learn More
1,2-Bis(2-aminophenoxy)-ethane-N,N,N'N'-tetraacetic acid, 97%
CAS: 85233-19-8 | C22H24N2O10 | 476.44 g/mol
$103.64 - $103.64
Identifiants chimiques
| CAS | 85233-19-8 |
|---|---|
| Molecular Formula | C22H24N2O10 |
| Molecular Weight (g/mol) | 476.44 |
| MDL Number | MFCD00036255 |
| InChI Key | FTEDXVNDVHYDQW-UHFFFAOYSA-N |
| Synonym | bapta, bapeta, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid, ca ii-bapta, 1,2-bis-2-aminophenoxy ethane n,n,n',n'-tetraacetic acid, unii-k22ddw77c0, 1,2-bis o-aminophenoxy ethane-n,n,n',n'-tetraacetic acid, bapta tetrasodium salt hydrate, glycine, n,n'-1,2-ethanediylbis oxy-2,1-phenylene bis n-carboxymethyl, 2,2',2,2'-ethane-1,2-diylbis oxy-2,1-phenylenenitrilo tetraacetic acid |
| PubChem CID | 104751 |
| ChEBI | CHEBI:60888 |
| IUPAC Name | 2-{[2-(2-{2-[bis(carboxymethyl)amino]phenoxy}ethoxy)phenyl](carboxymethyl)amino}acetic acid |
| SMILES | OC(=O)CN(CC(O)=O)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC(O)=O)CC(O)=O |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC123311000
|
Thermo Scientific Chemicals
123311000 |
100 mg | Glass bottle |
chaque for $103.64
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagentIdentifiants chimiques
| 85233-19-8 | |
| 476.44 | |
| FTEDXVNDVHYDQW-UHFFFAOYSA-N | |
| 104751 | |
| 2-{[2-(2-{2-[bis(carboxymethyl)amino]phenoxy}ethoxy)phenyl](carboxymethyl)amino}acetic acid |
| C22H24N2O10 | |
| MFCD00036255 | |
| bapta, bapeta, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid, ca ii-bapta, 1,2-bis-2-aminophenoxy ethane n,n,n',n'-tetraacetic acid, unii-k22ddw77c0, 1,2-bis o-aminophenoxy ethane-n,n,n',n'-tetraacetic acid, bapta tetrasodium salt hydrate, glycine, n,n'-1,2-ethanediylbis oxy-2,1-phenylene bis n-carboxymethyl, 2,2',2,2'-ethane-1,2-diylbis oxy-2,1-phenylenenitrilo tetraacetic acid | |
| CHEBI:60888 | |
| OC(=O)CN(CC(O)=O)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC(O)=O)CC(O)=O |
Spécifications
| 85233-19-8 | |
| 100.0 | |
| White | |
| 97% | |
| C22H24N2O10 | |
| 100 mg | |
| bapta, bapeta, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid, ca ii-bapta, 1,2-bis-2-aminophenoxy ethane n,n,n',n'-tetraacetic acid, unii-k22ddw77c0, 1,2-bis o-aminophenoxy ethane-n,n,n',n'-tetraacetic acid, bapta tetrasodium salt hydrate, glycine, n,n'-1,2-ethanediylbis oxy-2,1-phenylene bis n-carboxymethyl, 2,2',2,2'-ethane-1,2-diylbis oxy-2,1-phenylenenitrilo tetraacetic acid | |
| FTEDXVNDVHYDQW-UHFFFAOYSA-N | |
| 2-{[2-(2-{2-[bis(carboxymethyl)amino]phenoxy}ethoxy)phenyl](carboxymethyl)amino}acetic acid | |
| 104751 | |
| 476.44 | |
| Powder |
| 96.0 | |
| 177.0°C to 179.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00036255 | |
| 15, 952 | |
| Solubility in water: soluble. | |
| OC(=O)CN(CC(O)=O)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC(O)=O)CC(O)=O | |
| 476.44 | |
| CHEBI:60888 | |
| 97% | |
| 1,2-Bis(2-aminophenoxy)-ethane-N,N,N'N'-tetraacetic acid |
Sécurité et manipulation
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO – Research Use Only