Learn More
1-(1-Naphthyl)ethylamine, 98%
CAS: 42882-31-5 | C12H13N | 171.24 g/mol
$93.05 - $93.05
Identifiants chimiques
| CAS | 42882-31-5 |
|---|---|
| Molecular Formula | C12H13N |
| Molecular Weight (g/mol) | 171.24 |
| InChI Key | RTCUCQWIICFPOD-UHFFFAOYSA-N |
| Synonym | +/-1-1-naphthyl ethylamine, 1-1-naphthyl ethylamine, 1-naphthalen-1-yl ethanamine, dl-1-1-naphthyl ethylamine, 1-1-naphthyl ethanamine, 1-naphthalen-1-yl ethan-1-amine, +/--1-1-naphthyl ethylamine, 1-1-napthyl ethylamine, 1-naphthalen-1-yl-ethylamine, r-+-1-1-naphthyl ethylamine |
| PubChem CID | 98089 |
| IUPAC Name | 1-naphthalen-1-ylethanamine |
| SMILES | CC(C1=CC=CC2=CC=CC=C21)N |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AC450570050
|
Thermo Scientific Chemicals
450570050 |
5 g |
chaque for $93.05
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 42882-31-5 | |
| 171.24 | |
| +/-1-1-naphthyl ethylamine, 1-1-naphthyl ethylamine, 1-naphthalen-1-yl ethanamine, dl-1-1-naphthyl ethylamine, 1-1-naphthyl ethanamine, 1-naphthalen-1-yl ethan-1-amine, +/--1-1-naphthyl ethylamine, 1-1-napthyl ethylamine, 1-naphthalen-1-yl-ethylamine, r-+-1-1-naphthyl ethylamine | |
| 1-naphthalen-1-ylethanamine |
| C12H13N | |
| RTCUCQWIICFPOD-UHFFFAOYSA-N | |
| 98089 | |
| CC(C1=CC=CC2=CC=CC=C21)N |
Spécifications
| 42882-31-5 | |
| 156°C (15.0 mmHg) | |
| 98% | |
| C12H13N | |
| 1.063 | |
| Solubility in water: insoluble | |
| CC(C1=CC=CC2=CC=CC=C21)N | |
| 171.24 | |
| 171.24 | |
| 1-(1-Naphthyl)ethylamine |
| 1.0630g/mL | |
| >110°C | |
| Glass bottle | |
| 5 g | |
| +/-1-1-naphthyl ethylamine, 1-1-naphthyl ethylamine, 1-naphthalen-1-yl ethanamine, dl-1-1-naphthyl ethylamine, 1-1-naphthyl ethanamine, 1-naphthalen-1-yl ethan-1-amine, +/--1-1-naphthyl ethylamine, 1-1-napthyl ethylamine, 1-naphthalen-1-yl-ethylamine, r-+-1-1-naphthyl ethylamine | |
| RTCUCQWIICFPOD-UHFFFAOYSA-N | |
| 1-naphthalen-1-ylethanamine | |
| 98089 | |
| 98% |
Sécurité et manipulation
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO – Research Use Only