Learn how one site is manufacturing plastics that help customers like you develop life-saving therapies.
Thermo Scientific Matrix Barcoded Storage Tubes
Thermo Scientific™ Matrix™ Barcoded Storage Tubes provide robust sample protection and tracking efficiency. They’re 100% leak tested and designed for automated systems, including liquid handling, decappers, and other automated platforms. Permanent, high-contrast 2D, 1D linear, and human readable barcodes—that won’t fall off in freezers or liquid nitrogen tanks—offer three levels of identification.
Manufacturing Critical Products
Tubes, Racks, and Storage Options
Find a range of tube sizes, rack configurations, barcode styles, capping systems, caps, and more to meet your specific sample security and automation needs.
Accessories
Develop comprehensive systems for your specific sample storage and traceability requirements with compatible decappers, barcode readers, and sealers.
Catalog Number 4531009
Catalog Number 14754635
Catalog Number 4531012
Catalog Number 14754540