Learn More
Thermo Scientific™ VisionMate™ ST Single-Tube 2D Barcode Reader
Description
Scan 2D barcoded tubes into virtually any application with the compact Thermo Scientific™ VisionMate™ ST Single-Tube 2D Barcode Reader. Plug the unit into an available USB port and allow the drive to automatically install. Hold any Thermo Scientific™ 2D barcoded storage tube over the scanning window and decode the barcode instantly.
- Instant read time
- Simple to install and use
- For individual sample retrieval
- Export via comma-separated variable file, Microsoft™ Excel™, XML, COM port, ODBC or TCP socket
Specifications
Specifications
| Description | VisionMate ST 2-D Bar Code Reader |
| Data Transfer Method | Comma-separated variable file, Microsoft™ Excel™ |
| Quantity | 1/Pk., 1/Cs. |
| Type | Barcode Reader |
| Connection Type | USB |