missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Capit-All™ Screw Cap Tube Capper/Decapper

Catalog No. p-4531012
Change view
Click to view available options
Decapping Format:
48-Channel
96-Channel
For Use With (Equipment):
Matrix Screw Top Caps
Nunc Screw Top Caps
Speed:
∼10 sec./Cycle (Cap or Decap)
∼10 sec./Rack
∼15 sec./Cycle (Cap or Decap)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Decapping Format For Use With (Equipment) Speed
14754637 96-Channel Matrix Screw Top Caps ∼10 sec./Rack
14754638 96-Channel Matrix Screw Top Caps ∼15 sec./Cycle (Cap or Decap)
14754640 48-Channel Nunc Screw Top Caps ∼10 sec./Cycle (Cap or Decap)
14754641 48-Channel Nunc Screw Top Caps ∼15 sec./Cycle (Cap or Decap)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14754637 Supplier Thermo Scientific™ Supplier No. 4111MAT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Maintain sample integrity with a hands-free method of capping and decapping tubes quickly and easily.

Maintain sample integrity with a hands-free method of capping and decapping tubes quickly and easily with the Thermo Scientific™ Capit-All™ Screw Cap Tube Capper/Decapper. Designed for benchtop or integrated use, Capit-All instruments cap and decap 24-, 48-, and 96- format Thermo Scientific™ Matrix™ and Thermo Scientific™ Nunc™ ScrewTop tubes with precision and ease. All instruments cap tubes to optimal torque.

Enhanced Productivity

  • Increase throughput by capping and decapping an entire rack at a time in approximately 10 seconds
  • Automatically remove and replace caps on tubes of differing heights, even in the same rack, as well as tubes not properly seated in racks
  • Completely replace manual capping and decapping methods

Superior Sample Protection

  • Caps all tubes in a rack to the optimal torque level, maintaining sample integrity and preventing sample loss during storage
  • Safety features ensure racks are loaded in the correct orientation and that caps are not cross-threaded or overtightened
  • Drip tray prevents cross contamination of samples during addition and removal of tube racks
  • Internal construction ensures debris is not introduced into uncapped tubes

Ease of Use

  • The simple, four-button user interface enables laboratory personnel of any level to operate the instrument
  • Easily integrated into automated systems
  • Easily maintained and able to process up to a million tubes before requiring preventative maintenance

Specifications

Compatible Storage Tubes Matrix Style Screw Top Capped Tubes
Decapping Format 96-Channel
Description Capper/Decapper
Dimensions (L x W x H) 14.2 x 12.9 x 22 in. (32.8 x 36.0 x 56.0 cm)
Electrical Requirements 110 to 240VAC 50/60Hz
Net Weight (English) 97 lb.
Net Weight (Metric) 45 kg
No. per Case 1
No. per Pack 1
Operation Manual via Push Buttons
Voltage 110/240 V
Type Capper/Decapper
For Use With (Equipment) Matrix Screw Top Caps
Frequency 50/60 Hz
Speed ∼10 sec./Rack
Show More Show Less