missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Matrix™ ScrewTop Tube Caps, Cap Trays and Cap Inserts
GREENER_CHOICE

Catalog No. p-6155280
Change view
Click to view available options
Color:
Black
Blue
Colorless
Gray
Green
Orange
Pink
Purple
Red
White
Yellow
Product Type:
Screw Cap
Screw Cap Tray
Screw Cap Tray, Empty
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
14754668 Black Screw Cap
14754671 Orange Screw Cap
14754672 Pink Screw Cap
14754680 Gray Screw Cap Tray
14754681 Pink Screw Cap Tray
14754667 Colorless Screw Cap
14754674 Red Screw Cap
14754676 Yellow Screw Cap
14754670 Green Screw Cap
14754675 White Screw Cap
14754673 Purple Screw Cap
14754698 Yellow Screw Cap Tray, Empty
14754677 Colorless Screw Cap Tray
14754683 Red Screw Cap Tray
14754685 Yellow Screw Cap Tray
14754678 Blue Screw Cap Tray
14754679 Green Screw Cap Tray
14754684 White Screw Cap Tray
14754682 Purple Screw Cap Tray
14754669 Blue Screw Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Catalog No. 14754668 Supplier Thermo Scientific™ Supplier No. 4470BLK

Simplify sample storage with these color-coded caps and inserts, available racked in trays or in bulk to suit a variety of applications.

Simplify sample storage with these color-coded caps and inserts. Thermo Scientific™ Matrix™ ScrewTop Tube caps and inserts are available racked in trays or in bulk to suit a variety of applications.

Superior Cap Design

  • Gasketed caps ensure critical sample protection over the storage lifetime
  • Soft gasket compresses to ensure a proper seal, preventing contamination from outside the tube
  • Cap material: Virgin class VI medical grade polypropylene with virgin class VI medical grade silicone O-ring
  • ScrewTop caps are compatible with storage down to -196ºC

Cap Options Simplify Sample Identification

  • Cap trays feature standard microplate format compatible with automated equipment
  • Caps are available in seven color options to enable sample identification
  • Color inserts snap into cap for a secure fit; inserts are removable
  • Inserts are available in six colors

All caps and inserts are supplied free from DNA, RNase, DNase, endotoxins and cytotoxins

Specifications

Product Type Screw Cap
Material Polypropylene
Color Black
Packaging Bulk, 500 Caps
Sterility Sterile
For Use With (Equipment) ScrewTop Tube
Fit for Purpose Sterile and PCR Ready