thermo-scientific-shakers-rockers-and-rotators-d
Rockers
compact-digital-rocker
Thermo Scientific Compact Digital Rocker
Ensure gentle sample agitation with our compact, adjustable angle, digital display unit.
Features:
  • PID control ensures consistent and smooth rocking motion
  • Digital speed control: 5 to 100RPM
  • Easy-to-adjust tilt angle: 7° to 13°
  • Compact design is ideal for placing in incubators (non-CO2), chambers and refrigerators (up to 60°C)
  • Timer range: continuous or 1 minute to 99 hours, 59 minutes
  • Includes dimpled mat (tray, silicone pad, two securing bands)
Applications
Designed for time-dependent and low-dependent sensitive applications such as ELISA, protein synthesis, staining/de-staining gels, and antibody staining.
Buy Compact Digital Rocker Now
test-tube-rockers
Thermo Scientific Vari-Mix and Speci-Mix Test Tube Rockers
Offer a choice of fixed - or adjustable-angle models.
Features:
  • All models hold 10 to 30mm diameter tubes and are ideal for blood collection tubes; Choose from fixed or adjustable speeds
  • Reversible pad provides one side (with lip to prevent sliding) for capped tubes 110mm (4.33in.) long; other side for longer tubes
  • Autoclavable white silicone rubber pad enhances viewing
  • Can be used in a noncondensing incubator or coldroom with ambient operating conditions of 4° to 40°C (39.2° to 104°F) and 20 to 80% RH noncondensing
Buy Vari-Mix and Speci-Mix Test Tube Rockers Now
Rotators
bottle-tube-roller
Thermo Scientific CO2 Resistant Digital Compact Bottle/Tube Roller
Maximize valuable bench space with our compact, adjustable-speed unit, capable of rolling multiple tube and bottle sizes.
Features
  • Adjustable speed accommodates a variety of applications
  • Digital speed control from 1 -80 RPM
  • Small footprint ideal for use in a CO2 incubator
  • Anti-corrosive and wear-resistant construction enables use in humid and CO2 environments
  • Adjustable rollers accommodate a variety of tube/bottle sizes
  • Stacking kit allows you to stack units two high, saving valuable bench space
  • Optional remote control facilitates operating unit while in incubator and add timer function
  • Includes pack of O-rings for tubes. O-rings ensure consistent smooth rolling when using tubes
Applications
Cell Culture | Hybridization Assays | Liquid Mixing
Buy Digital Compact Bottle/Tube Rollers Now
tube-revolver
Thermo Scientific Digital Tube Revolver
Increase your lab's efficiency with our variable-speed tube revolver. Its small footprint, flexible design and simple display help increase flexibility and effectiveness.
Features
  • Interchangeable rotisseries accommodate different tube sizes for maximum flexibility. Each paddle can be adjusted from 0-90 degrees
  • Achieve more effective mixing with "oscillating" mode. Oscillation will occur at every 40 degrees in the rotation cycle
  • Small footprint
  • Plug-and-play simplicity
  • Maintenance-free motor, quiet operation
Standard Unit Includes:
  • 90-691-36 — 10mL/15mL paddle, Quantity 2
  • 90-691-37 — 5mL/7mL paddle, Quantity 2
  • 90-691-38 — 1.5mL/2mL paddle, Quantity 2
  • 90-691-40 — 50mL paddle, Quantity 1
Applications
Molecular Biology | Clinical Biochemistry
Buy Tube Revolvers Now
compact-digital-waving-rotator
Thermo Scientific Compact Digital Waving Rotator
Achieve maximum sample mixing efficiency with the adjustable orbital waving digital-display rotator.
Features:
  • PID control ensures consistent and smooth rocking motion
  • Digital Speed Control: 8 to100rpm
  • Easy to adjust tilt angle: 0° to 13°
  • Compact design ideal for placing in incubators (non-CO2), chambers and refrigerators (up to 60°C)
  • Timer range: continuous or 1 minute to 99 hours 59 minutes
  • Includes dimpled mat (tray, silicone pad, two securing bands)
  • When no angle is used, the Waving Rotator can be used as a slow speed orbital shaker
Applications
Designed for time-dependent and low-dependent sensitive applications such as ELISA, protein synthesis, staining/de-staining gels and antibody staining.
Buy Compact Digital Waving Rotators Now
culture-rotator
Thermo Scientific Cel-Gro Tissue Culture Rotator
Available in a single or dual drum configuration.
Features
  • Single-drum as an adjustable rotating angle and speed, while the dual-drum has a fixed rotating angle and speed
  • Rotator operates in standard incubators for cultivation under controlled temperature conditions
  • Powder-coated stainless-steel outer body construction is durable and corrosion-resistant
  • Drums can be lifted off the rotor without interrupting rotator operation
  • Adjusting screw on the base varies elevation angle
  • Adjustable 5° rotating angle above horizontal to 90°
  • Adjustable speed control from 12 to 70rpm
  • Drum required with purchase, sizes vary based on tube size requirements:
    • 142 x 17mm tube diameter
    • 76 x 26mm tube diameter
    • 60 x 30mm tube diameter
Buy Cel-Gro Tissue Culture Rotators Now
compact-digital-mini-rotator
Thermo Scientific Compact Digital Mini Rotator
Realize maximum flexibility and high quality with the easy-to-use Thermo Scientificâ„¢ Compact Digital Mini Rotator/Shaker, which provides smooth, quiet orbital mixing for beakers, flasks, tubes, dishes and more.
Features
  • PID control ensures consistent and smooth orbital motion
  • Digital Speed Control from 50 to 300rpm
  • Timer Range: Continuous or 1min to 99hrs 59min.
  • Compact design ideal for placing in incubators (non-CO2), chambers and refrigerators (up to 60°C). 
  • Choose from 5 different top options to customize unit to meet exact application needs.
  • Display both current and set RPM simultaneously.
Applications
  • Bacteria and fungi culturing
  • Hybridization assays
  • Gel staining/de-staining
Buy Compact Digital Mini Rotator Now
compact-digital-microplate-shaker
Thermo Scientificâ„¢ Compact Digital Microplate Shaker
Harness the efficiency of our microplate shaker, capable of shaking up to four microplates in one run.
Features
  • PID control ensures consistent shaking results
  • Digital display for easy viewing
  • Compact design is ideal for placing in incubators (non-CO2), chambers, and refrigerators (up to 60°C)
  • Timer Range: Continuous or 1min to 99hrs 59min
  • Automatic Overload Shutdown: 4.4kg / 9.7lbs
  • Small 3mm orbital motion, ideal mixing small volumes
  • Tube rack accessories make this unit extremely vesitile
  • Includes 4 place microplate top standard
Applications
  • Any application requiring precise and accurate shaking of microplates: immunoassay, screening applications, magnetic bead applications
Buy Compact Digital Microplate Shaker Now
Mixers
Thermo Scientific LP Vortex Mixer
Choose to continuously mix or pulse samples with our low-profile vortex mixer.
Features:
  • Low-profile design with small footprint
  • Run in continuous or pulsing mode
  • Speed control: 0 to 300rpm
  • Automatic overload shut off
  • Wide variety of optional accessories to accommodate different vessel types
Applications
Any application that requires intense vortex mixing in a tube, plate or glass vessel.
Buy LP Vortex Mixers Now
Special Offers
Sorry, there are currently no special offers at this time.
Check back throughout the year to find ways to save or view all of our promotions.
Resources