Thermo Scientific Biological Safety Cabinets
The Difference Is the Design
With Thermo Scientific™ biological safety cabinets, you never have to think about user and sample protection. Designed with Thermo Scientific™ SmartFlow™ technology, the cabinets include dual-DC motors to automatically balance the cabinet inflow and downflow air velocities in real time—even as the filters load with contaminants. Plus, the Digital Airflow Verification (DAVe) alarm signals any out-of-spec conditions for added assurance. Combined with the reliability, ergonomic features, and energy efficiency of Thermo Scientific brand products, the ideal choice is also the one you can trust completely. And not just on day one.
Innovations in Containment and Efficiency
A biological safety cabinet's primary function is to provide cleanliness and containment. Features available on Thermo Scientific biological safety cabinets include:
- SmartFlow dual-motor design to automate airflow compensation and balance
- Digital Airflow Verification (DAVe) to alert users to excessive flow restriction
- Night Set-Back mode that reduces airflow to 30% when window is closed
- A 75% reduction in energy use and a longer motor and filter life than traditional AC motor cabinets
- Reduced noise level enhances user focus
- 10° sloped front for exceptional ergonomics
- Spacious work area with comfortable armrest
- Increased light intensity for brighter work environment
- SmartClean front window design for easy cleaning
- Intuitive control panel for at-a-glance monitoring
- SmartPort organizes tubing and cables with convenient side access
- Programmable UV lights extends bulb life and saves energy
- The built-in WiFi system* and touchscreen are ready for cloud-based capabilities like remote equipment monitoring, secure data storage, collaboration tools, and more
- Glass side walls and LED lighting make for a brighter workspace with better visibility
- SmartClean Plus motorized, hinged front window design enables easycleaning
- Dual DC motors power the SmartFlow Plus airflow compensation system while delivering better energy efficiency and quieter operation
- The crossbeam UV light system features an exclusive self-adjusting exposure capability to enable reproducible germicidal effectiveness with every use
- The SmartPort allows you to easily pass tubing and wiring through the sidewall to accommodate application needs
- Work surfaces can be customized to suit your application and work style
- The built-in WiFi system* and touchscreen are ready for cloud-based capabilities like remote equipment monitoring, secure data storage, collaboration tools, and more
View More Thermo Scientific Biological Safety Cabinets
*Availability of connectivity-enabled models depends on region. Please contact your sales representative to learn more.
-
Eppendorf Scientific IncLimited Time
Save up to 45% when you buy qualifying Eppendorf liquid handling products.
Buy Now Expires: 2025-06-30 -
METTLER TOLEDOLimited Time
Combine your purchase of qualifying METTLER TOLEDO balances or analytical instruments to earn more.
Buy Now Expires: 2025-07-31 -
Greiner Bio-OneLimited Time
Buy three cases of Greiner Bio-One Serological Pipets and get an extra case at no additional cost.
Buy Now Expires: 2025-07-15 -
BiotixLimited Time
Buy three qualifying cases of BioTix Pipette Tips and receive a fourth case of tips.
Buy Now Expires: 2025-08-31 -
Cytiva
When you buy one qualifying Cytiva filtration product, you’ll get extra products at no additional cost.
Buy Now Expires: 2025-08-31 -
Cytiva
Buy one Cytiva product and get an identical product at no additional cost.
Buy Now Expires: 2025-08-31 -
Gibco,Life Technologies Gibco Process Liquids
Buy four qualifying Gibco Buffers and get one extra buffer at no additional cost.
Buy Now Expires: 2025-10-31 -
MP Biomedicals Inc
Buy three identical MP Biomedicals whole blood fractionation products and get a fourth identical product.
Buy Now Expires: 2025-09-30 -
Ricca Chemical Company
Buy two Ricca Chemical Titrants and get an extra titrant of equal or lesser value at no additional cost.
Buy Now Expires: 2025-10-31 -
MilliporeSigmaLimited Time
Buy three qualifying packs of MilliporeSigma Filtration Units and get an extra pack at no additional cost.
Buy Now Expires: 2025-07-31 -
MilliporeSigma
Buy a MilliporeSigma Scepter 3.0 Handheld Automated Cell Counter Kit and get a pack of sensors.
Buy Now Expires: 2025-10-31 -
Thermo ScientificLimited Time
Buy one Thermo Scientific Orion Pro Star Meter and get three Thermo Scientific Orion Buffers.
Buy Now Expires: 2025-07-31 -
Buchi Corporation
Buy a BUCHI Freeze Dryer and Pump and get a Mobile Trolley and your choice of a Drying Chamber or Manifold.
Buy Now Expires: 2025-10-31 -
Thermo Fisher ScientificLimited Time
Buy a Thermo Scientific Heratherm Chamber and get three extra shelves for your product.
Buy Now Expires: 2025-06-30 -
Corning IncorporatedLimited Time
Buy five or more bottles Corning Fetal Bovine Sera and get double the quantity of the same serum.
Buy Now Expires: 2025-07-15
Thermo Scientific Herasafe 2030i Class II A2 Biological Safety Cabinet
The Herasafe 2030i is equipped with “smart” self-monitoring safety features, an intuitive graphic touchscreen, and built-in connectivity to preserve essential performance data.
Learn More About the Thermo Scientific Herasafe 2030i Class II A2 Biological Safety Cabinet
Thermo Scientific 1300 Class II A2 Biological Safety Cabinet
This biological safety cabinet helps protect users and keep them comfortable with a proprietary airflow design, ergonomic features, and energy efficiency for operational cost savings.
Learn More About the Thermo Scientific 1300 Class II A2 Biological Safety Cabinet
Thermo Scientific Biological Safety Cabinets Airflow Alarms Flyer
Why are airflow alarms an important consideration for Biological Safety Cabinets (BSCs)?
Thermo Scientific Biological Safety Cabinets DC Motor Flyer
Why should you select a Class II Biological Safety Cabinet (BSC) with a DC motor over one with an AC motor?
Thermo Scientific Biological Safety Cabinets Dual Blower Flyer
Why is a dual blower system better than a single blower system?
Thermo Scientific Biological Safety Cabinets Filter Life Flyer
How do I extend the filter life of my biological safety cabinet (BSC)?
More Resources
Thermo Scientific Biological Safety Cabinets Operating Costs Data Sheet
Managing groups of biological safety cabinets (BSCs) requires an optimization strategy that addresses both performance and cost.