Thermo Scientific Orbital Shakers
- Featured Products
- Resources
Trust your samples to the outstanding line of Thermo Scientific™ orbital shakers.
Thermo Scientific™ MaxQ™ basic and advanced shakers contain a triple-eccentric drive mechanism to accommodate heavier loads and provide uniform shaking. Designed to ensure 24/7 operation, MaxQ shakers also feature maintenance-free brushless DC motors.
Thermo Scientific™ Solaris™ orbital shakers set the new standard for reliability and technical innovation. Ruggedly built for demanding use, they combine a new ergonomic design with key feature enhancements that save you time, protect your process and drive your science forward.
Each shaker comes with a two-year warranty on parts, a five-year warranty on labor, and a 10-year warranty on the drive mechanism.
Top Product Categories:
CO2 Resistant Shakers
Get reliable, around-the-clock operation ideally suited to keep your cells alive and flourishing within your working environment.
Open Air
Get the choice and flexibility you need from a variety of basic and advanced benchtop open air shaker models. Separately available platforms for some models allow you to increase capacity simply by changing the platform.
Incubated/Refrigerated
Orbital shakers come in a wide range of temperature capabilities to meet specific growth requirements. Upgrade to any of our advanced incubated/refrigerated shakers to suit your application's shaking needs.
Stackable
Save lab space and complete large-capacity applications with our floor and stackable orbital shakers. Combine incubated and refrigerated shaker capabilities with easy-to-use, ergonomic controls to maximize application flexibility and production.
High Performance
When you need a reliable solution for sensitive cell cultures requiring protection from contamination, choose from the MaxQ high-performance line of shakers.
Resources
- Filename
- thermo-scientific-shaker-tradein-flyer.pdf
- Size
- 270 KB
- Format
- application/pdf
- Filename
- thermo-scientific-max-q-shakers.pdf
- Size
- 8 MB
- Format
- application/pdf
- Filename
- thermo-scientific-shake-up-world-science-flier.pdf
- Size
- 871 KB
- Format
- application/pdf
- Filename
- orbital-shaker-benchmarks-best-practices.pdf
- Size
- 2 MB
- Format
- application/pdf