Thermo Scientific Nalgene Reusable Plastic Labware
Discoveries can happen any day
Featured Products
Make Your Lab Safer and Smarter
Thermo Scientific™ Nalgene™ plastic labware is lighter than glass and doesn’t shatter, so it’s safer and easier to use. Since it’s more affordable than glass and durable enough to last for years, you can do even more with your budget.
Browse these products:
Bottles
Shapes and Materials for Every Application
Nalgene bottles are made from a wide variety of plastics to fit virtually any laboratory application. Choose from polypropylene copolymer (PPCO), low- and high-density polyethylene (LDPE, HDPE), polyethylene terephthalate (PETG or copolymer G), polycarbonate (PC) and fluoropolymer (FEP-PFA).
narrow-mouth-bottles-17-0931
Narrow-Mouth Bottles
Nalgene narrow-mouth bottles provide more controlled pouring than wide-mouth versions. Choose narrow-mouth lab-quality bottles, large lab bottles, low-cost and lightweight economy bottles, and space-saving square options in PPCO, FEP and HDPE.
Browse Now
wide-mouth-bottles-17-0931
Wide-Mouth Bottles
The wide-mouth design of Nalgene laboratory bottles makes them easier to fill and use with solid, semi-solid and viscous liquid materials. Options include lab-quality bottles, economy bottles, and space-saving square and rectangular bottles in PMP, LDPE and HDPE.
Browse Now
wash-bottles-17-0931
Wash Bottles
Nalgene plastic wash bottles are sturdily constructed and are always straight-shooters. They are available in multiple resins and styles that include top-dispensing, safety-labeled or UnitaryTM wash bottles that feature a unique vent system that won’t drip.
Browse Now
dropper-bottles-17-0931
Dropper Bottles
Nalgene dropper bottles are available in a variety of volumes and formats. Dropper bottles with colored screw caps provide controlled, measured drop-wise dispensing, and bottles with tethered caps are our most popular. We also offer bottles with our bulb-transfer pipette-style cap assembly forfaster dispensing or to access harder-to-reach places. For an autoclavable dropper bottle, choose PPCO or opt for a fluoropolymer (FEP) with a captive spout cap for dispensing more aggressive chemicals or high-purity reagents.
Browse Now
General Plasticware
Durable, Affordable Labware
Substitute Nalgene plastic labware for glass when mixing or manipulating to reduce the likelihood of breakage and keep users safer.
beakers-burets-cylinders-17-0931
Beakers, Burets and Cylinders
Nalgene beakers are available in Griffin low-form style or with convenient handles and are made with a variety of resins. The plastic burets meet Class B (ASTM E287) accuracy, and liquids measured in Nalgene burets or cylinders don't form a meniscus, which makes reading them easier.
Browse Now
dessicators-17-0931
Desiccators
Nalgene desiccators are lighter and easier to move around the lab. They can spare you from injury and may even save you up to 60% as compared to glass desiccators.
Browse Now
flasks-funnels-17-0931
Flasks and Funnels
Nalgene Erlenmeyer flasks, funnels and separatory funnels easily replace their glass counterparts that chip and break. Volumetric flasks are individually hand calibrated, engraved and inked with a volume fill line to provide precision and Class B (ASTM E228/ISO 384) accuracy.
Browse Now
tubing-17-0931
Tubing
Nalgene tubing comes in various formulations and sizes, including clear PVC tubing (including Tygon™ equivalents), food-grade tubing that complies with USP Class VI and USDA regulations, flexible braided tubing for higher-pressure applications, and other polyurethane, LDPE, PP, PFA and FEP tubing.
Browse Now
Carboys and Containers
Sizes and Styles for Every Lab
We offer a versatile selection of carboys and jugs made from virgin plastics with minimal additives in round, rectangular and lowboy shapes up to 50L in capacity. Choose between options with or without spigots and with molded or stainless-steel carry handles.
carboys-jugs-17-0931
Carboys and Jugs
Nalgene round carboys are suitable for collecting and dispensing distilled water, acids and reagents. Nalgene low-density polyethylene or polypropylene jugs feature a large carrying handle and excellent chemical resistance.
Browse Now
buckets-pails-17-0931
Buckets and Pails
Store and transport dry and solid materials, specimens, and general lab supplies with our range of buckets and other containers, available in a variety of materials.
Browse Now
pans-trays-baskets-17-0931
Pans, Trays and Baskets
Repeatedly autoclave, dry, and sterilize instruments and labware with Nalgene heavy-duty pans, trays, and baskets.
Browse Now
jars-17-0931
Jars
Made from the highest-quality laboratory-grade plastic materials for dependably low extractables, Nalgene jars offer easy access and viewing of contents.
Browse Now
Lab Safety and Organization
Stay Clean and Neat
Keep your staff safe and your lab organized with our full range of safety and organization products. We offer everything from bench protectors and lab notebooks to splash shields and waste disposal devices.
bench-protectors-17-0931
Bench Protectors
Nalgene surface protectors are cushioned to minimize breakage and feature a plastic backing to prevent liquid breakthrough.
Browse Now
racks-carriers-totes-17-0931
Racks, Carriers and Totes
Transport bottles and other lab essentials safely and easily using Thermo Scientific racks, totes, and carriers. 
Browse Now
safety-shields-17-0931
Safety Shields
Protect against splashes, aerosols and flying debris while maintaining a clear field of vision with Nalgene safety shields.
Browse Now
waste-disposal-17-0931
Waste Disposal
Safely dispose of biohazardous waste as well as solvents and chemicals with Nalgene waste containers.
Browse Now
All Products
Filtered Search Results
Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.
Narrow Results
Narrow Results
  • (85)
  • (22)
  • (1)
  • (21)
  • (560)
  • (16)
  • (559)
  • (4)
  • (1)
  • (21)
  • (1)
  • (4)
  • (2)
  • (85)
  • (96)
  • (1)
  • (13)
  • (44)
  • (81)
  • (1)
  • (39)
  • (1)
  • (7)
  • (1)
  • (74)
  • (2)
  • (4)
  • (2)
  • (2)
  • (49)
  • (5)
  • (1)
  • (22)
  • (1)
  • (6)
  • (6)
  • (1)
  • (1)
  • (9)
  • (1)
  • (32)
  • (1)
  • (1)
  • (1)
  • (34)
  • (1)
  • (2)
  • (9)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (8)
  • (2)
  • (1)
  • (1)
  • (1)
  • (10)
  • (1)
  • (7)
  • (1)
  • (1)
  • (4)
  • (45)
  • (2)
  • (1)
  • (1)
  • (19)
  • (4)
  • (1)
  • (1)
  • (11)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (9)
  • (1)
  • (70)
  • (1)
  • (22)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (6)
  • (7)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)
  • (18)
  • (1)
  • (4)
  • (2)
  • (2)
  • (16)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (4)
  • (9)
  • (2)
  • (2)
  • (1)
  • (35)
  • (12)
  • (3)
  • (8)
  • (2)
  • (1)
  • (24)
  • (2)
  • (2)
  • (18)
  • (9)
  • (1)
  • (18)
  • (11)
  • (9)
  • (14)
  • (12)
  • (14)
  • (4)
  • (48)
  • (2)
  • (2)
  • (22)
  • (1)
  • (13)
  • (2)
  • (18)
  • (17)
  • (14)
  • (1)
  • (15)
  • (24)
  • (11)
  • (43)
  • (11)
  • (5)
  • (13)
  • (159)
  • (17)
  • (42)
  • (11)
  • (6)
  • (54)
  • (68)
  • (163)
  • (1)
  • (36)
  • (2)
  • (3)
  • (4)
  • (3)
  • (6)
  • (9)
  • (1)
  • (10)
  • (18)
  • (1)
  • (60)
  • (1)
  • (6)
  • (22)
  • (1)
  • (1)
  • (1)
  • (37)
  • (27)
  • (1)
  • (1)
  • (3)
  • (3)
  • (29)
  • (1)
  • (144)
  • (94)
  • (146)
  • (72)
  • (4)
  • (1)
  • (2)
  • (12)
  • (3)
  • (8)
  • (19)
  • (1)
  • (4)
  • (81)
  • (6)
  • (8)
  • (9)
  • (10)
  • (6)
  • (2)
  • (2)
  • (35)
Special Offers
Resources
More Resources