Thermo Scientific Orbital Shakers
Thermo Scientific Orbital Shakers
Featured Products
Trust your samples to the outstanding line of Thermo Scientificâ„¢ orbital shakers.
Thermo Scientificâ„¢ MaxQâ„¢ basic and advanced shakers contain a triple-eccentric drive mechanism to accommodate heavier loads and provide uniform shaking. Designed to ensure 24/7 operation, MaxQ shakers also feature maintenance-free brushless DC motors.
Thermo Scientificâ„¢ Solarisâ„¢ orbital shakers set the new standard for reliability and technical innovation. Ruggedly built for demanding use, they combine a new ergonomic design with key feature enhancements that save you time, protect your process and drive your science forward.
Each shaker comes with a two-year warranty on parts, a five-year warranty on labor, and a 10-year warranty on the drive mechanism.
Top Product Categories:
Upgrade and Save
Upgrade and Save
Trade up to a new Thermo Scientific Solaris or MaxQ shaker and improve your lab’s efficiency.
Learn More
CO2 Resistant Shakers
Get reliable, around-the-clock operation ideally suited to keep your cells alive and flourishing within your working environment.
CO2 Resistant Shakers
CO2 Resistant Shakers
• Orbital shaking through magnetic power eliminates the need to replace belts
• Small footprint ideal for use in a CO2 incubator
• Specially treated mechanical components protect up to 20% CO2 and 95% humidity conditions
Browse Now
MaxQ 416 HP
MaxQ 416 HP
• Low heat 24V DC motor with moisture- and CO2-resistant electronics provides continuous maintenance-free operation on the benchtop or in a CO2 chamber
• Triple-counter-balanced mechanism for optimum handling of full or unbalanced loads, regardless of vessel placement; completely eliminates "walking"
• Units operate in temperature range of 4°C to 40°C and humidity conditions from 50% to 80% non-condensing enabling them to be used in a variety of environments
Browse Now
Open Air
Get the choice and flexibility you need from a variety of basic and advanced benchtop open air shaker models. Separately available platforms for some models allow you to increase capacity simply by changing the platform.
Solaris 2000/4000
Solaris 2000/4000
• Completely sealed system that helps to reduce any internal corrosion or contamination, making these units suitable for a variety of incubators (non-CO2), warm rooms, environmental chambers, and refrigerators
• Programmable features allow for up to 99 unique saved programs for easy setup and reproducibility
• Full-color touchscreen display allows you to clearly view speed and run times concurrently, even from a distance
Browse Now
Incubated/Refrigerated
Orbital shakers come in a wide range of temperature capabilities to meet specific growth requirements. Upgrade to any of our advanced incubated/refrigerated shakers to suit your application's shaking needs.
MaxQ HP Console Shakers
MaxQ HP Console Shakers
• Shakers feature HEPA filtration for maximum protection from contamination
• Directed airflow system for optimum temperature uniformity and ideal growth conditions
• Front drain system makes the chamber easy to access and clean
Browse Now
Stackable
Save lab space and complete large-capacity applications with our floor and stackable orbital shakers. Combine incubated and refrigerated shaker capabilities with easy-to-use, ergonomic controls to maximize application flexibility and production.
MaxQ 6000
MaxQ 6000
  • Units can be stacked two high on the floor to save valuable bench space
  • Two adjustable-height shelves provide added storage
  • Large viewing window and internal light offer sample visibility
  • Available in incubated or refrigerated models
MaxQ 8000
MaxQ 8000
  • Units can be stacked three high on the floor to save valuable space and still be run at full speed, even when stacked
  • HEPA filter evenly distributes air inside the chamber and reduces cross-contamination
  • Ergonomically designed fold-down door and slide-out platform
  • Includes stacking kit
IntelliStack
IntelliStack
  • Units can be stacked three high to save space
  • Combines the functionality of an incubator and a shaker
  • Large, easy-to-use 10 in. color touchscreen display
  • Available in 25 mm or 50 mm orbits
  • Available as a CO2 Incubator Shaker or Incubator Shaker
High Performance
When you need a reliable solution for sensitive cell cultures requiring protection from contamination, choose from the MaxQ high-performance line of shakers.
MaxQ 435 HP, 436 HP, and 481 HP
MaxQ 435 HP, 436 HP, and 481 HP
• Operating temperatures: Incubated: 5°C above ambient to 60°C (41°F to 140°F) or Incubated/Refrigerated: 4°C above ambient to 60°C (39.2° to 140°F) in 0.1°C increments with ±0.1°C control
• HEPA filtration for maximum protection from contamination
Browse Now
All Products
Narrow Results
Narrow Results
  • (154)
  • (14)
  • (30)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (4)
  • (4)
  • (1)
  • (8)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
Special Offers
Sorry, there are currently no special offers at this time.
Check back throughout the year to find ways to save or view all of our promotions.
Resources