accessibility menu, dialog, popup

UserName

CAS RN 982-57-0

IUPAC Name: sodium 4-[2-(2,2-dichloroacetamido)-3-hydroxy-3-(4-nitrophenyl)propoxy]-4-oxobutanoate
Synonyms: sodium 4-[2-(2,2-dichloroacetamido)-3-hydroxy-3-(4-nitrophenyl)propoxy]-4-oxobutanoate
Molecular Weight (g/mol): 445.18
Molecular Formula: C15H15Cl2N2NaO8
InChi Key: RPLOPBHEZLFENN-UHFFFAOYNA-M
SMILES: [Na+].OC(C(COC(=O)CCC([O-])=O)NC(=O)C(Cl)Cl)C1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)