missing translation for 'onlineSavingsMsg'
Learn More

Chloramphenicol Sodium Succinate, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022847
Change view
Click to view available options
Quantity:
1500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022847 1500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022847 Supplier Merck Emd Millipore Supplier No. BP1061

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

Chemical Name or Material Pharmacopeia & Metrological Institutes Standards
CAS 982-57-0
Molecular Formula C15H15Cl2N2NaO8
Linear Formula C15H15Cl2N2O8Na
MDL Number MFCD00079025
Quantity 1500 mg
Synonym Chloramphenicol succinate sodium salt; Chloramphenicol alpha-succinate
Formula Weight 445.18
Grade British Pharmacopoeia (BP) Reference Standard