missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chloramphenicol sodium succinate, 98-102%

Catalog No. AC459530250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459530250 25 g
AC459530050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459530250 Supplier Thermo Scientific Chemicals Supplier No. 459530250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 982-57-0 | C15H15Cl2N2NaO8 | 445.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 982-57-0
Molecular Formula C15H15Cl2N2NaO8
Molecular Weight (g/mol) 445.18
InChI Key RPLOPBHEZLFENN-HTMVYDOJSA-M
Synonym succinic acid chloramphenicol sodium
PubChem CID 73940259
IUPAC Name sodium;4-[(2R,3R)-2-[(2,2-dichloroacetyl)amino]-3-hydroxy-3-(4-nitrophenyl)propoxy]-4-oxobutanoate
SMILES C1=CC(=CC=C1C(C(COC(=O)CCC(=O)[O-])NC(=O)C(Cl)Cl)O)[N+](=O)[O-].[Na+]

Specifications

Infrared Spectrum Authentic
Assay Percent Range 98.0 to 102% on dried substance
Quantity 25 g
Formula Weight 445.18
Percent Purity 98 to 102%
Chemical Name or Material Chloramphenicol sodium succinate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned.
EINECSNumber 213-568-1

RUO – Research Use Only