accessibility menu, dialog, popup

UserName

H3CCH3OOCH3CH2

CAS RN 97-86-9

IUPAC Name: 2-methylpropyl 2-methylprop-2-enoate
Synonyms: isobutyl methacrylate
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: RUMACXVDVNRZJZ-UHFFFAOYSA-N
SMILES: CC(C)COC(=O)C(C)=C
Boiling Point 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)