missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl methacrylate, 99.5+%, stab. with 10ppm 4-methoxyphenol

Catalog No. AA41948K7
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41948K7 4 L
AA41948AK 250 mL
AA41948K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA41948K7 Supplier Thermo Scientific Chemicals Supplier No. 041948K7

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 97-86-9 | C8H14O2 | 142.20 g/mol

Plastic industry: used in the manufacture of containing active hydroxyl acrylic resin. Coatings with epoxy resin, diisocyanate and melamine formaldehyde resin configuration used for preparing a two-component coating. In oil industry as additives for lubricating oil washing. In electronics industry used as electron microscope to take off the water. Textile industry used in the manufacture of adhesive fabric. Used as chemical reagents in analytical chemistry. In addition, also used for water immiscible embedding agent, used in the synthesis of medical polymer materials, thermosetting coatings and adhesives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97-86-9
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00008931,MFCD00084435
InChI Key RUMACXVDVNRZJZ-UHFFFAOYSA-N
Synonym isobutyl methacrylate, 2-propenoic acid, 2-methyl-, 2-methylpropyl ester, methacrylic acid, isobutyl ester, isobutyl 2-methyl-2-propenoate, 2-methylpropyl methacrylate, isobutyl alpha-methacrylate, isobutyl alpha-methylacrylate, unii-v11534uyz0, ccris 4829, isobutylester kyseliny methakrylove
PubChem CID 7352
IUPAC Name 2-methylpropyl 2-methylprop-2-enoate
SMILES CC(C)COC(=O)C(C)=C

Specifications

Density 0.889 g/mL
Boiling Point 155°C
Flash Point 46°C (115°F)
Quantity 4 L
UN Number UN2283
Beilstein 1747595
Solubility Information Soluble in water at 20°C,2g/L
Formula Weight 142.2
Percent Purity ≥99.5%
Physical Form Liquid
Chemical Name or Material Isobutyl methacrylate, Stabilized with 10ppm 4-methoxyphenol
Show More Show Less
Hazard Category H226-H315-H317-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H317-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ISOBUTYL METHACRYLATE, STABILIZED
EINECSNumber 202-613-0
RTECSNumber OZ4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only