accessibility menu, dialog, popup

UserName

CAS RN 97-74-5

IUPAC Name: N,N-dimethyl[(dimethylcarbamothioyl)sulfanyl]carbothioamide
Synonyms: monex bis(dimethylthiocarbamoyl) sulfide (tmtm) thiuram monosulphides
Molecular Weight (g/mol): 208.36
Molecular Formula: C6H12N2S3
InChi Key: REQPQFUJGGOFQL-UHFFFAOYSA-N
SMILES: CN(C)C(=S)SC(=S)N(C)C
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)