missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylthiuram monosulfide, 97%

Catalog No. AAA1725922
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1725922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1725922 Supplier Thermo Scientific Chemicals Supplier No. A1725922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 97-74-5 | C6H12N2S3 | 208.356 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97-74-5
Molecular Formula C6H12N2S3
Molecular Weight (g/mol) 208.356
MDL Number MFCD00014870
InChI Key REQPQFUJGGOFQL-UHFFFAOYSA-N
Synonym tetramethylthiuram monosulfide, bis dimethylthiocarbamoyl sulfide, monothiuram, unads, mono-thiurad, tmtm, thiuram mm, aceto tmtm, cyuram ms, ekagom tm
PubChem CID 7347
IUPAC Name dimethylcarbamothioyl N,N-dimethylcarbamodithioate
SMILES CN(C)C(=S)SC(=S)N(C)C

Specifications

Melting Point 106°C to 110°C
Density 1.37
Quantity 100 g
Beilstein 1775650
Formula Weight 208.37
Percent Purity 97%
Chemical Name or Material Tetramethylthiuram monosulfide
Hazard Category H302-H317
Hazard Statement GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 202-605-7
RTECSNumber WQ1750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only