accessibility menu, dialog, popup

UserName

H3CCH3(S)NHCH3OOHO

CAS RN 96-81-1

IUPAC Name: (2S)-2-acetamido-3-methylbutanoic acid
Synonyms: L-valine, N-acetyl-
Molecular Weight (g/mol): 159.19
Molecular Formula: C7H13NO3
InChi Key: IHYJTAOFMMMOPX-LURJTMIESA-N
SMILES: CC(C)[C@H](NC(C)=O)C(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)