missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-valine 98.0+%, TCI America™

Catalog No. A23665G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23665G 5 g
A236625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23665G Supplier TCI America Supplier No. A23665G
Only null left

Chemical Identifiers

CAS 96-81-1
Molecular Formula C7H13NO3
Molecular Weight (g/mol) 159.19
MDL Number MFCD00066066
InChI Key IHYJTAOFMMMOPX-LURJTMIESA-N
Synonym n-acetyl-l-valine, ac-val-oh, l-valine, n-acetyl, s-2-acetamido-3-methylbutanoic acid, acetyl-l-valine, n-acetylvaline, 2s-2-acetamido-3-methylbutanoic acid, unii-u83p7h9hv3, acetylvaline, valine, n-acetyl-, l
PubChem CID 66789
IUPAC Name (2S)-2-acetamido-3-methylbutanoic acid
SMILES CC(C)[C@H](NC(C)=O)C(O)=O

Specifications

Melting Point 165°C
Color White
Quantity 5 g
Formula Weight 159.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-L-valine