missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-valine, 98%, Thermo Scientific Chemicals

Catalog No. AAH6694314
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6694314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6694314 Supplier Thermo Scientific Chemicals Supplier No. H6694314
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96-81-1
Molecular Formula C7H13NO3
Molecular Weight (g/mol) 159.19
MDL Number MFCD00066066
InChI Key IHYJTAOFMMMOPX-LURJTMIESA-N
Synonym n-acetyl-l-valine, ac-val-oh, l-valine, n-acetyl, s-2-acetamido-3-methylbutanoic acid, acetyl-l-valine, n-acetylvaline, 2s-2-acetamido-3-methylbutanoic acid, unii-u83p7h9hv3, acetylvaline, valine, n-acetyl-, l
PubChem CID 66789
IUPAC Name (2S)-2-acetamido-3-methylbutanoic acid
SMILES CC(C)[C@H](NC(C)=O)C(O)=O

Specifications

Melting Point 163°C to 167°C
Color White
Beilstein 1723837
Quantity 25 g
Formula Weight 159.19
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-L-valine
EINECSNumber 202-537-8
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only