accessibility menu, dialog, popup

UserName

CAS RN 955964-73-5

H3CCH3NBrBr

CAS RN 955964-73-5

IUPAC Name: 2,7-dibromo-9-(heptadecan-9-yl)-9H-carbazole
Synonyms: 2,7-dibromo-9-(heptadecan-9-yl)carbazole
Molecular Weight (g/mol): 563.46
Molecular Formula: C29H41Br2N
InChi Key: HRUNMSCVUSIWQM-UHFFFAOYSA-N
SMILES: CCCCCCCCC(CCCCCCCC)N1C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results