missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,7-Dibromo-9-(9-heptadecyl)carbazole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 72°C |
| Color | White-Yellow |
| Quantity | 200 mg |
| Formula Weight | 563.46 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,7-Dibromo-9-(9-heptadecyl)carbazole |
Safety and Handling
| TSCA | No |