accessibility menu, dialog, popup

UserName

CAS RN 95407-69-5

CH3OH3CH3CNOH3CCH3

CAS RN 95407-69-5

IUPAC Name: 4-methoxy-2,2,6,6-tetramethylpiperidin-1-olate
Synonyms: 4-methoxy-2,2,6,6-tetramethylpiperidin-1-olate
Molecular Weight (g/mol): 186.28
Molecular Formula: C10H20NO2
InChi Key: BGXWUNHAMWRFOR-UHFFFAOYSA-N
SMILES: COC1CC(C)(C)N([O-])C(C)(C)C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results