missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 98.0+%, TCI America™

Catalog No. M11975G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11975G 5 g
M11971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11975G Supplier TCI America Supplier No. M11975G
Only null left

Chemical Identifiers

CAS 95407-69-5
Molecular Formula C10H20NO2
MDL Number MFCD00270334
Synonym 4-Methoxy-TEMPO

Specifications

Melting Point 43°C
Quantity 5 g
Formula Weight 186.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical [Catalyst for Oxidation]