accessibility menu, dialog, popup

UserName

CH3SOOO

CAS RN 953-91-3

IUPAC Name: cyclohexyl 4-methylbenzene-1-sulfonate
Synonyms: cyclohexyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 254.34
Molecular Formula: C13H18O3S
InChi Key: OHHPZPDQZMUTCA-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OC1CCCCC1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)