missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl p-toluenesulfonate, 97%

Catalog No. AAL1795214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1795214 25 g
AAL1795206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1795214 Supplier Thermo Scientific Chemicals Supplier No. L1795214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 953-91-3 | C13H18O3S | 254.344 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 953-91-3
Molecular Formula C13H18O3S
Molecular Weight (g/mol) 254.344
MDL Number MFCD00014291
InChI Key OHHPZPDQZMUTCA-UHFFFAOYSA-N
Synonym cyclohexyl p-toluenesulfonate, cyclohexyl tosylate, cyclohexyl toluenesulfonate, cyclohexylp-toluenesulfonate, cyclohexyl p-toluenesulphonate, benzenesulfonic acid, 4-methyl-, cyclohexyl ester, p-toluenesulfonic acid, cyclohexyl ester, cyclohexyl-p-toluene sulfonate, benzenesulfonic acid, cyclohexyl ester, acmc-20aj06
PubChem CID 13722
IUPAC Name cyclohexyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC2CCCCC2

Specifications

Melting Point 45°C to 46°C
Quantity 25 g
Beilstein 2217391
Formula Weight 254.35
Percent Purity 97%
Chemical Name or Material Cyclohexyl p-toluenesulfonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-468-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only