missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. C23635G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23635G 5 g
C236325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23635G Supplier TCI America Supplier No. C23635G

Chemical Identifiers

CAS 953-91-3
Molecular Formula C13H18O3S
Molecular Weight (g/mol) 254.344
MDL Number MFCD00014291
InChI Key OHHPZPDQZMUTCA-UHFFFAOYSA-N
Synonym cyclohexyl p-toluenesulfonate, cyclohexyl tosylate, cyclohexyl toluenesulfonate, cyclohexylp-toluenesulfonate, cyclohexyl p-toluenesulphonate, benzenesulfonic acid, 4-methyl-, cyclohexyl ester, p-toluenesulfonic acid, cyclohexyl ester, cyclohexyl-p-toluene sulfonate, benzenesulfonic acid, cyclohexyl ester, acmc-20aj06
PubChem CID 13722
IUPAC Name cyclohexyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC2CCCCC2

Specifications

Melting Point 45°C
Color White-Yellow
Quantity 5 g
Formula Weight 254.34
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cyclohexyl p-Toluenesulfonate