accessibility menu, dialog, popup

UserName

NH2HO

CAS RN 95-55-6

IUPAC Name: 2-aminophenol
Synonyms: 2-aminophenol benzofur GG zoba 3GA 2-hydroxyanaline CI oxidation base 17 nako yellow ga paradone olive green B monosodium salt 2-aminophenol pelagol 3GA nako yellow 3GA
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: CDAWCLOXVUBKRW-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1O
Molecular Weight (g/mol) 
  • (7)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (6)
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (4)
  • (3)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (6)
  • (7)
  • (3)
  • (1)
  • (3)
  • (6)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
  • (3)
  • (3)