missing translation for 'onlineSavingsMsg'
Learn More

2-Aminophénol 98,0+%, TCI America™

Catalog No. A0335500G
Change view
Click to view available options
Quantité:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
A0335500G 500 g
A033525G 25 g
A0335100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0335500G Supplier TCI America Supplier No. A0335500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 95-55-6
Formule moléculaire C6H7NO
Poids moléculaire (g/mol) 109.128
Numéro MDL MFCD00007690
Clé InChI CDAWCLOXVUBKRW-UHFFFAOYSA-N
Synonyme o-aminophenol, o-hydroxyaniline, 2-hydroxyaniline, phenol, 2-amino, 2-amino-1-hydroxybenzene, fouramine op, benzofur gg, pelagol grey gg, pelagol 3ga, nako yellow 3ga
PubChem CID 5801
ChEBI CHEBI:18112
Nom de l’IUPAC 2-aminophénol
SOURIRES C1=CC=C(C(=C1)N)O

Specifications

Nom ou substance chimique 2-Aminophenol
Point de fusion 175 °C
Quantité 500 g
Numéro UN 2512
Poids de formule 109.13
Pourcentage de pureté ≥98,0% (T)
Forme physique Poudre cristalline
EINECSNumber (3)-0675
RTECSNumber SJ4950000
TSCA Oui