accessibility menu, dialog, popup

UserName

NBr

CAS RN 94847-10-6

IUPAC Name: 9-(6-bromohexyl)-9H-carbazole
Synonyms: 9-(6-bromohexyl)carbazole
Molecular Weight (g/mol): 330.27
Molecular Formula: C18H20BrN
InChi Key: HFNNZEMVTJAKRC-UHFFFAOYSA-N
SMILES: BrCCCCCCN1C2=C(C=CC=C2)C2=C1C=CC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results