missing translation for 'onlineSavingsMsg'
Learn More

9-(6-Bromohexyl)-9H-carbazole 97.0+%, TCI America™

Catalog No. B5548200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5548200MG 200 mg
B55481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5548200MG Supplier TCI America Supplier No. B5548200MG
Only null left

Chemical Identifiers

CAS 94847-10-6
Molecular Formula C18H20BrN
Molecular Weight (g/mol) 330.27
MDL Number MFCD30562129
InChI Key HFNNZEMVTJAKRC-UHFFFAOYSA-N
PubChem CID 11738992
IUPAC Name 9-(6-bromohexyl)-9H-carbazole
SMILES BrCCCCCCN1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 60°C
Color White-Yellow
Quantity 200 mg
Formula Weight 330.27
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(6-Bromohexyl)-9H-carbazole