accessibility menu, dialog, popup

UserName

CAS RN 945-51-7

IUPAC Name: (benzenesulfinyl)benzene
Synonyms: diphenyl sulfoxide
Molecular Weight (g/mol): 202.27
Molecular Formula: C12H10OS
InChi Key: JJHHIJFTHRNPIK-UHFFFAOYSA-N
SMILES: O=S(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (5)