missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Sulfoxide 99.0+%, TCI America™

Catalog No. D10025G
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10025G 5 g
D100225G 25 g
D1002100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D10025G Supplier TCI America Supplier No. D10025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 945-51-7
Molecular Formula C12H10OS
Molecular Weight (g/mol) 202.271
MDL Number MFCD00002085
InChI Key JJHHIJFTHRNPIK-UHFFFAOYSA-N
Synonym diphenyl sulfoxide, phenyl sulfoxide, sulfinyldibenzene, phenylsulfinylbenzene, diphenyl sulphoxide, sulfoxide, diphenyl, diphenylsulfoxide, benzene, 1,1'-sulfinylbis, 1,1'-sulfinyldibenzene, phenylsulfinyl benzene
PubChem CID 13679
IUPAC Name benzenesulfinylbenzene
SMILES C1=CC=C(C=C1)S(=O)C2=CC=CC=C2

Specifications

Melting Point 71°C
Color White
Boiling Point 198°C
Quantity 5 g
Formula Weight 202.27
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl Sulfoxide
RTECSNumber DA9185000
TSCA Yes