accessibility menu, dialog, popup

UserName

H3COONH2

CAS RN 94-25-7

IUPAC Name: butyl 4-aminobenzoate
Synonyms: butamben butyl keloform abbott brand of butamben picrate butamben picrate preparation scuroform scuroforme butamben (product) butyl aminobenzoate (substance) N-butyl p-aminobenzoate butyl paba
Molecular Weight (g/mol): 193.25
Molecular Formula: C11H15NO2
InChi Key: IUWVALYLNVXWKX-UHFFFAOYSA-N
SMILES: CCCCOC(=O)C1=CC=C(N)C=C1
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)