missing translation for 'onlineSavingsMsg'
Learn More

Butyl 4-Aminobenzoate 99.0+%, TCI America™

Catalog No. A027025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A027025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A027025G Supplier TCI America Supplier No. A027025G

Chemical Identifiers

CAS 94-25-7
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.246
MDL Number MFCD00017112
InChI Key IUWVALYLNVXWKX-UHFFFAOYSA-N
Synonym butamben, butesin, butylcaine, butyl aminobenzoate, butesine, butoform, planoform, scuroform, scuroforme, butyl keloform
PubChem CID 2482
ChEBI CHEBI:3231
IUPAC Name butyl 4-aminobenzoate
SMILES CCCCOC(=O)C1=CC=C(C=C1)N

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 174°C
Quantity 25 g
Formula Weight 193.25
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Butyl 4-Aminobenzoate
EINECSNumber (3)-1443
RTECSNumber DG1530000
TSCA Yes