accessibility menu, dialog, popup

UserName

H3CNHONHSOOCl

CAS RN 94-20-2

IUPAC Name: 1-(4-chlorobenzenesulfonyl)-3-propylurea
Synonyms: chlorpropamide diamel ex novo-propamide diabaril diabenese diabeneza chlorpropamide (inn) chlorpropamide bp/ usp insulase chlorporpamide
Molecular Weight (g/mol): 276.74
Molecular Formula: C10H13ClN2O3S
InChi Key: RKWGIWYCVPQPMF-UHFFFAOYSA-N
SMILES: CCCNC(=O)NS(=O)(=O)C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Grade 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)