missing translation for 'onlineSavingsMsg'
Learn More

Chlorpropamide Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015944
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015944 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015944 Supplier Merck Emd Millipore Supplier No. PHR12841G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Chlorpropamide belongs to the sulfonylurea class of drugs and is found to be clinically effective in the treatment of Diabetes mellitus (DM). Its mode of action involves the stimulation of insulin release from the β-cells of the pancreas in response to a glucose load. It helps in increasing the number of insulin receptors, thereby enhancing the effect of insulin on the liver and promotes peripheral glucose utilization. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 94-20-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C10H13 ClN2O3 S
MDL Number MFCD00079004
Synonym 1-(p-Chlorobenzenesulfonyl)-3-propylurea
Type Certified Reference Material
Quantity 1 g
Formula Weight 276.74
Grade Certified Reference Material