missing translation for 'onlineSavingsMsg'
Learn More

Chlorpropamide, 97%, MilliporeSigma™ Supelco™

Catalog No. 111015943
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015943 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015943 Supplier Merck Emd Millipore Supplier No. C129025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chlorpropamide is a drug that belongs to the sulfonylurea family. It is an effective candidate in the treatment of diabetes insipidus.

Featured Industries: Pharmaceutical (small molecule)

Specifications

Chemical Name or Material Chlorpropamide
CAS 94-20-2
Physical Form Neat
Linear Formula C10H13 ClN2O3 S
Technique HPLC
MDL Number MFCD00079004
Synonym 1-(p-Chlorobenzenesulfonyl)-3-propylurea
Type Analytical Standard
Molecular Weight (g/mol) 276.74
Quantity 25 g
Formula Weight 276.74
Percent Purity ≥97%
Grade Analytical Standard
Show More Show Less