missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenylsulfonyl)-3-propylurea 99.0+%, TCI America™

Catalog No. C122025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C122025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C122025G Supplier TCI America Supplier No. C122025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 94-20-2
Molecular Formula C10H13ClN2O3S
Molecular Weight (g/mol) 276.735
MDL Number MFCD00079004
InChI Key RKWGIWYCVPQPMF-UHFFFAOYSA-N
Synonym chlorpropamide, chloropropamide, diabinese, chlorpropamid, diabenese, glucamide, meldian, chlorodiabina, chloronase, diabeneza
PubChem CID 2727
ChEBI CHEBI:3650
IUPAC Name 1-(4-chlorophenyl)sulfonyl-3-propylurea
SMILES CCCNC(=O)NS(=O)(=O)C1=CC=C(C=C1)Cl

Specifications

Melting Point 128°C
Color White
Quantity 25 g
Formula Weight 276.74
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Chlorophenylsulfonyl)-3-propylurea
RTECSNumber YS6650000
TSCA No