accessibility menu, dialog, popup

UserName

CAS RN 94-13-3

IUPAC Name: propyl 4-hydroxybenzoate
Synonyms: propylparaben betacine P propyl hydroxybenzoate propyl aseptoform caswell no. 714 p-oxybenzoesaeurepropylester pulvis conservans P-hydroxy propyl benzoate propylparasept fema number 2951
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: QELSKZZBTMNZEB-UHFFFAOYSA-N
SMILES: CCCOC(=O)C1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (4)
  • (8)
  • (2)
  • (7)
Percent Purity 
  • (3)
  • (4)
  • (2)
  • (4)
  • (4)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
Physical Form 
  • (8)
  • (8)
Grade 
  • (2)
  • (4)
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (15)
  • (10)
  • (2)
  • (2)
  • (8)
  • (4)
  • (7)
  • (4)
  • (8)
  • (2)
  • (7)
  • (3)
  • (4)
  • (2)
  • (4)
  • (4)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (8)
  • (8)
  • (2)
  • (4)
  • (4)
  • (2)