missing translation for 'onlineSavingsMsg'
Learn More

Propyl 4-Hydroxybenzoate 99.0+%, TCI America™

Catalog No. P19555G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19555G 5 g
P195525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P19555G Supplier TCI America Supplier No. P19555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 94-13-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00002354
InChI Key QELSKZZBTMNZEB-UHFFFAOYSA-N
Synonym propylparaben, propyl paraben, 4-hydroxybenzoic acid propyl ester, propyl p-hydroxybenzoate, nipasol, nipazol, propyl butex, betacide p, propyl parahydroxybenzoate, propylparasept
PubChem CID 7175
ChEBI CHEBI:32063
IUPAC Name propyl 4-hydroxybenzoate
SMILES CCCOC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 98°C
Color White
Quantity 5 g
Formula Weight 180.20
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Propyl 4-Hydroxybenzoate [for Biochemical Research]
EINECSNumber (3)-1585
RTECSNumber DH2800000
TSCA Yes