missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Propylparaben, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| Product Line | TraceCERT |
| CAS | 94-13-3 |
| Linear Formula | HOC6H4 CO2 CH2 CH2 CH3 |
| MDL Number | MFCD00002354 |
| Melting Point | 95°C to 98°C |
| Synonym | Propyl 4-hydroxybenzoate; 4-Hydroxybenzoic acid propyl ester; Propyl paraben |
| Type | Certified Reference Material |
| Quantity | 100 mg |
| Formula Weight | 180.20 |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC