missing translation for 'onlineSavingsMsg'
Learn More

Propylparaben Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015942
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015942 10 g
111015941 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015942 Supplier Merck Emd Millipore Supplier No. PHR101010G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Propylparaben is the benzoate ester, widely used as a preservative in personal care products. Featured Industries: Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

CAS 94-13-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula HOC6H4 CO2 CH2 CH2 CH3
MDL Number MFCD00002354
Melting Point 95°C to 98°C
Synonym Propyl 4-hydroxybenzoate; 4-Hydroxybenzoic acid propyl ester; Propyl paraben
Type Certified Reference Material
Quantity 10 g
Formula Weight 180.20
Grade Certified Reference Material
Show More Show Less