accessibility menu, dialog, popup

UserName

NaCH3H3CNF(R)OH(S)OHOO(E)

CAS RN 93957-55-2

IUPAC Name: sodium (3S,5R,6E)-7-[3-(4-fluorophenyl)-1-(propan-2-yl)-1H-indol-2-yl]-3,5-dihydroxyhept-6-enoate
Synonyms: sodium (3S,5R,6E)-7-[3-(4-fluorophenyl)-1-isopropylindol-2-yl]-3,5-dihydroxyhept-6-enoate
Molecular Weight (g/mol): 433.46
Molecular Formula: C24H25FNNaO4
InChi Key: ZGGHKIMDNBDHJB-RPQBTBOMSA-M
SMILES: [Na+].CC(C)N1C(\C=C\[C@H](O)C[C@H](O)CC([O-])=O)=C(C2=CC=CC=C12)C1=CC=C(F)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)