missing translation for 'onlineSavingsMsg'
Learn More

Fluvastatin sodium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015937
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015937 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015937 Supplier Merck Emd Millipore Supplier No. PHR16201G
Pharmaceutical (small molecule)

Specifications

Packaging Ampule
CAS 93957-55-2
Linear Formula C24H25 FNNaO4
MDL Number MFCD08063391
Synonym (3 R,5 S,6 E)-rel-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1 H-indol-2-yl]-3,5-dihydroxy-6-heptenoic acid monosodium salt
Type Certified Reference Material
Quantity 1 g
Formula Weight 433.45
Grade Certified Reference Material