missing translation for 'onlineSavingsMsg'
Learn More

Fluvastatin Sodium Salt Hydrate 98.0+%, TCI America™

Catalog No. F08205G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08205G 5 g
F08201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08205G Supplier TCI America Supplier No. F08205G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 93957-55-2
Molecular Formula C24H25FNNaO4
Molecular Weight (g/mol) 433.46
MDL Number MFCD00929076
InChI Key ZGGHKIMDNBDHJB-RPQBTBOMSA-M
Synonym fluvastatin sodium, sodium 3s,5r-7-3-4-fluorophenyl-1-propan-2-yl-1h-indol-2-yl-3,5-dihydroxyhept-6-enoate
PubChem CID 66587062
IUPAC Name sodium (3S,5R,6E)-7-[3-(4-fluorophenyl)-1-(propan-2-yl)-1H-indol-2-yl]-3,5-dihydroxyhept-6-enoate
SMILES [Na+].CC(C)N1C(\C=C\[C@H](O)C[C@H](O)CC([O-])=O)=C(C2=CC=CC=C12)C1=CC=C(F)C=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 433.46
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Fluvastatin Sodium Salt Hydrate
RTECSNumber MJ9675100
TSCA No
Recommended Storage Refrigerator