accessibility menu, dialog, popup

UserName

CH3OOH3CH3C

CAS RN 924-50-5

IUPAC Name: methyl 3-methylbut-2-enoate
Synonyms: methyl 3-methylbut-2-enoate
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: FZIBCCGGICGWBP-UHFFFAOYSA-N
SMILES: COC(=O)C=C(C)C
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)